Skip to Content
Merck
All Photos(1)

Key Documents

66158

Supelco

3-(4-Methylbenzylidene)camphor

analytical standard

Synonym(s):

1,7,7-Trimethyl-3-(4-methylbenzylidene)-norbornan-2-one, 1,7,7-Trimethyl-3-[(4-methylphenyl)methylene]-bicyclo[2.2.1]heptan-2-one, 4-MBC, 4-Methylbenzylidene camphor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O
CAS Number:
Molecular Weight:
254.37
Beilstein:
9213949
EC Number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cc1ccc(cc1)\C=C2\[C@@H]3CC[C@](C)(C2=O)C3(C)C

InChI

1S/C18H22O/c1-12-5-7-13(8-6-12)11-14-15-9-10-18(4,16(14)19)17(15,2)3/h5-8,11,15H,9-10H2,1-4H3/b14-11-/t15-,18+/m0/s1

InChI key

HEOCBCNFKCOKBX-IRELMECMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

372.2 °F

Flash Point(C)

189 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos E Rodríguez-Rodríguez et al.
Bioresource technology, 168, 180-189 (2014-03-04)
The use of Trametes versicolor has been partially successful in the removal of some pharmaceuticals from sewage sludge in laboratory-scale biopile systems. The application of two strategies for the re-inoculation of biomass was assessed during the fungal bioaugmentation of non-sterile
C Couteau et al.
International journal of pharmaceutics, 476(1-2), 160-163 (2014-10-01)
Given that previous studies have highlighted the anti-inflammatory nature, which is influenced by UV radiation, of a certain number of ultraviolet filters currently used in Europe, it seemed interesting to evaluate the persistence of this type of effect. The persistence
Kasper Skov et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 978-979, 83-88 (2014-12-23)
Blood plasma is a well-known body fluid often analyzed in studies on the effects of toxic compounds as physiological or chemical induced changes in the mammalian body are reflected in the plasma metabolome. Sample preparation prior to LC-MS based analysis
Marina G Pintado-Herrera et al.
Analytica chimica acta, 851, 1-13 (2014-12-03)
This work presents the development, optimization and validation of a multi-residue method for the simultaneous determination of 102 contaminants, including fragrances, UV filters, repellents, endocrine disruptors, biocides, polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), and several types of pesticides in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service