Skip to Content
Merck
All Photos(1)

Documents

633879

Supelco

1,1,1,3,3,3-Hexafluoro-2-(fluoromethoxy)propane-d3

98 atom % D, 97% (CP)

Synonym(s):

Fluoromethyl 1,1,1,3,3,3-hexafluoro-2-propyl ether-d3, Fluoromethyl 2,2,2-trifluoro-1-(trifluoromethyl)ethyl ether-d3, Sevoflurane-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CD(OCD2F)CF3
CAS Number:
Molecular Weight:
203.07
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

bp

59 °C (lit.)

density

1.567 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])(F)OC([2H])(C(F)(F)F)C(F)(F)F

InChI

1S/C4H3F7O/c5-1-12-2(3(6,7)8)4(9,10)11/h2H,1H2/i1D2,2D

InChI key

DFEYYRMXOJXZRJ-FUDHJZNOSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service