Skip to Content
Merck
All Photos(1)

Key Documents

63056

Sigma-Aldrich

Magnesiumanthracene tetrahydrofuran complex

technical, ~90% (NMR)

Synonym(s):

9,10-Dihydro-9,10-anthracendiyl-tris(THF)magnesium, Magnesium anthracene dianion

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Mg · 3C4H8O
CAS Number:
Molecular Weight:
418.85
Beilstein:
4240840
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

grade

technical

Assay

~90% (NMR)

mp

~200 °C (dec.) (lit.)

SMILES string

[Mg].C1CCOC1.C2CCOC2.C3CCOC3.[CH]4c5ccccc5[CH]c6ccccc46

Looking for similar products? Visit Product Comparison Guide

Other Notes

Used in a catalyst system for the hydrogenation of magnesium to magnesium hydride[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B. Bogdanovic et al.
Angewandte Chemie (International Edition in English), 92, 845-845 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service