Skip to Content
Merck
All Photos(1)

Key Documents

62710

Sigma-Aldrich

Lutetium(III) trifluoromethanesulfonate

purum, ≥98.0% (KT)

Synonym(s):

Lutetium(III) triflate, Trifluoromethanesulfonic acid lutetium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Lu
CAS Number:
Molecular Weight:
622.17
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

grade

purum

Assay

≥98.0% (KT)

SMILES string

[Lu+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Lu/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

NRMNRSCGHRWJAK-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Other Notes

Water-tolerant, reusable catalyst fostering many reactions; e.g. aldol reactions[1]; Enantioselective Diels-Alder reactions[2]; Starting material for preparing lutetium organometallic compounds[3]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Kobayashi et al.
Journal of the American Chemical Society, 116, 4083-4083 (1994)
H. Schumann et al.
Journal of Organometallic Chemistry, 430, 299-299 (1992)
S. Kobayashi et al.
The Journal of Organic Chemistry, 59, 3590-3590 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service