Skip to Content
Merck
All Photos(1)

Key Documents

54470

Sigma-Aldrich

1-(2-Hydroxyethyl)piperidine

purum, ≥97.0% (GC)

Synonym(s):

1-Piperidineethanol, 2-Piperidinoethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
Beilstein:
103390
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.4804 (lit.)
n20/D 1.481

bp

199-202 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

OCCN1CCCCC1

InChI

1S/C7H15NO/c9-7-6-8-4-2-1-3-5-8/h9H,1-7H2

InChI key

KZTWONRVIPPDKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service