Skip to Content
Merck
All Photos(1)

Key Documents

49734

Sigma-Aldrich

mono-Methyl glutarate

technical, ≥97.0% (T)

Synonym(s):

Monomethyl glutarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)3COOCH3
CAS Number:
Molecular Weight:
146.14
Beilstein:
1762381
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

grade

technical

Assay

≥97.0% (T)

refractive index

n20/D 1.438 (lit.)
n20/D 1.439

bp

150-151 °C/10 mmHg (lit.)

density

1.169 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCC(O)=O

InChI

1S/C6H10O4/c1-10-6(9)4-2-3-5(7)8/h2-4H2,1H3,(H,7,8)

InChI key

IBMRTYCHDPMBFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service