Skip to Content
Merck
All Photos(1)

Key Documents

47822

Sigma-Aldrich

D-Fructose 2,6-bisphosphate sodium salt

≥97.0% (TLC)

Synonym(s):

D-Fructose 2,6-diphosphate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NaO12P2
CAS Number:
Molecular Weight:
362.10
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥97.0% (TLC)

solubility

H2O: 50 mg/mL, clear, very faintly yellow

storage temp.

2-8°C

SMILES string

[Na+].OC[C@]1(O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]1O)OP(O)([O-])=O

Other Notes

Reported to be a stimulator of fructose-6-phosphate kinase

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Uyeda et al.
The Journal of biological chemistry, 256(16), 8394-8399 (1981-08-25)
The effect of natural "activation factor" and synthetic fructose-2,6-P2 on the allosteric kinetic properties of liver and muscle phosphofructokinases was investigated. Both synthetic and natural fructose-2,6-P2 show identical effects on the allosteric kinetic properties of both enzymes. Fructose-2,6-P2 counteracts inhibition
S J Pilkis et al.
The Journal of biological chemistry, 256(7), 3171-3174 (1981-04-10)
A new activator of rat liver phosphofructokinase was partially purified from rat hepatocyte extracts by DEAE-Sephadex chromatography. The activator, which eluted in the sugar diphosphate region, was sensitive to acid treatment but resistant to heating in alkali. Mild acid hydrolysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service