Skip to Content
Merck
All Photos(2)

Key Documents

45963

Supelco

2,4-Dinitroaniline

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3NH2
CAS Number:
Molecular Weight:
183.12
Beilstein:
982999
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

176-178 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H5N3O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H,7H2

InChI key

LXQOQPGNCGEELI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dinitroaniline, typically derived as a nitro-substituted compound of aromatic amines, is an isomer of dinitroaniline, which finds primary use as an intermediate in the manufacture of dyestuff, pharmaceutical products, pesticides, etc.[1]

Application

2,4-Dinitroaniline may be used as an analytical standard for the determination of the analyte in environmental water samples and wastewater samples by chromatographic techniques.[1][2][3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

435.2 °F - closed cup

Flash Point(C)

224 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojia Huang et al.
Journal of chromatography. A, 1216(20), 4354-4360 (2009-04-03)
A simple and sensitive method for the determination of polar aromatic amines (PAAs) was developed using stir bar sorptive extraction (SBSE) coupling to high-performance liquid chromatography. A hydrophilic poly(vinylimidazole-divinylbenzene) (VIDB) monolithic material was prepared and acted as SBSE coating. The
C Bolognesi et al.
Bollettino della Societa italiana di biologia sperimentale, 56(23), 2480-2485 (1980-12-15)
The DNA damage induced by in vivo administration of: 2,4-dinitroaniline, ortho-toluidine and para-toluidine, was determined using alkaline filter elution of DNA. The target organs for ultimate carcinogens produced in mice appear to be the liver and the kidney. The damage
José Raul Herance et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(22), 6491-6502 (2005-08-12)
Zeolites are suitable microporous hosts for positively charged organic species, but it is believed that they cannot adsorb organic anions. Pure Meisenheimer complex, derived from reduction of 2,4-dinitroaniline with NaBH4, was adsorbed inside faujasite cavities. Evidence for the internal incorporation
[Establishment of the MPEL for 2,4-dinitroaniline in water by an experimental computational method].
N Ia Mikhaĭlovskiĭ et al.
Gigiena i sanitariia, (10)(10), 15-18 (1982-10-01)
B Striepen et al.
The Journal of cell biology, 151(7), 1423-1434 (2001-01-03)
Apicomplexan parasites harbor a single nonphotosynthetic plastid, the apicoplast, which is essential for parasite survival. Exploiting Toxoplasma gondii as an accessible system for cell biological analysis and molecular genetic manipulation, we have studied how these parasites ensure that the plastid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service