Skip to Content
Merck
All Photos(1)

Key Documents

443115

Supelco

Filling solution for cyanide or sulfide electrode (10% KNO3, trace AgCl, KCl)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
PubChem Substance ID:

bp

100 °C/750 mmHg

density

1.06 g/mL at 25 °C

General description

Visit our Sensor Applications portal to learn more.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service