Skip to Content
Merck
All Photos(1)

Key Documents

41150

Sigma-Aldrich

2,2-Dimethyl-1,3,6,9,12-pentaoxa-2-silacyclotetradecane

technical, ≥90% (GC)

Synonym(s):

2,2-Dimethyl-2-sila-14-crown-5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22O5Si
CAS Number:
Molecular Weight:
250.36
Beilstein:
6387584
EC Number:
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

SMILES string

C[Si]1(C)OCCOCCOCCOCCO1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service