Skip to Content
Merck
All Photos(1)

Key Documents

39375

Sigma-Aldrich

Titanium(IV) chloride

≥99.0% (AT)

Synonym(s):

TTC, Titanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl4
CAS Number:
Molecular Weight:
189.68
EC Number:
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

vapor pressure

50 mmHg ( 55 °C)
9.6 mmHg ( 20 °C)

Assay

≥99.0% (AT)

form

liquid

bp

135-136 °C (lit.)

mp

−25 °C (lit.)

density

1.73 g/mL at 20 °C (lit.)

SMILES string

Cl[Ti](Cl)(Cl)Cl

InChI

1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

XJDNKRIXUMDJCW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service