Skip to Content
Merck
All Photos(1)

Key Documents

38770

Sigma-Aldrich

Methyl dimethoxyacetate

purum, ≥95.0% (GC)

Synonym(s):

Glyoxylic acid methyl ester dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CHCOOCH3
CAS Number:
Molecular Weight:
134.13
Beilstein:
1757582
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum

Assay

≥95.0% (GC)

refractive index

n20/D 1.405 (lit.)
n20/D 1.406

bp

67 °C/18 mmHg (lit.)

density

1.096 g/mL at 25 °C (lit.)

SMILES string

COC(OC)C(=O)OC

InChI

1S/C5H10O4/c1-7-4(6)5(8-2)9-3/h5H,1-3H3

InChI key

NZTCVGHPDWAALP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Protected glyoxylic acid; preparation of methyl glyoxylate[1]; Preparation of α-oxo esters by alkylation[2]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.M. Hook
Synthetic Communications, 14, 83-83 (1984)
F. Huet et al.
Synthesis, 33-33 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service