Skip to Content
Merck
All Photos(1)

Key Documents

37098

Supelco

Methoxychlor-d14 (bis(4-methoxyphenyl-d7))

PESTANAL®, analytical standard

Synonym(s):

1,1,1-Trichloro-2,2-bis-(p-methoxy-d3-phenyl-d4)ethane, 2,2-Bis(4-methoxy-d3-phenyl-d4)-1,1,1-trichloroethane, Methoxychlor-d14

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D14HCl3O2
CAS Number:
Molecular Weight:
359.73
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

[2H]C1=C([2H])C(C(C(Cl)(Cl)Cl)C2=C([2H])C([2H])=C(OC([2H])([2H])[2H])C([2H])=C2[2H])=C([2H])C([2H])=C1OC([2H])([2H])[2H]

InChI

1S/C16H15Cl3O2/c1-20-13-7-3-11(4-8-13)15(16(17,18)19)12-5-9-14(21-2)10-6-12/h3-10,15H,1-2H3/i1D3,2D3,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

IAKOZHOLGAGEJT-DDAUHAMOSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service