Skip to Content
Merck
All Photos(1)

Key Documents

34820

Sigma-Aldrich

Dibutyl phosphite

technical, ≥95% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2O)2P(O)H
CAS Number:
Molecular Weight:
194.21
Beilstein:
1099706
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

vapor pressure

<0.1 mmHg ( 20 °C)

grade

technical

Assay

≥95% (GC)

refractive index

n20/D 1.423 (lit.)
n20/D 1.424

bp

118-119 °C/11 mmHg (lit.)

density

0.995 g/mL at 25 °C (lit.)

SMILES string

CCCCO[PH](=O)OCCCC

InChI

1S/C8H19O3P/c1-3-5-7-10-12(9)11-8-6-4-2/h12H,3-8H2,1-2H3

InChI key

NFJPGAKRJKLOJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 Oral - STOT RE 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Immune system, Liver,Kidney, thymus,Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service