Skip to Content
Merck
All Photos(1)

Key Documents

34250

Sigma-Aldrich

1,8-Dibromooctane

purum, ≥97.0% (GC)

Synonym(s):

Octamethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)8Br
CAS Number:
Molecular Weight:
272.02
Beilstein:
1698114
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.498 (lit.)
n20/D 1.499

bp

270-272 °C (lit.)

mp

12-16 °C (lit.)

density

1.477 g/mL at 25 °C (lit.)

SMILES string

BrCCCCCCCCBr

InChI

1S/C8H16Br2/c9-7-5-3-1-2-4-6-8-10/h1-8H2

InChI key

DKEGCUDAFWNSSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Jiang et al.
Cell, 183(1), 258-268 (2020-08-30)
Plasmodium species, the causative agent of malaria, rely on glucose for energy supply during blood stage. Inhibition of glucose uptake thus represents a potential strategy for the development of antimalarial drugs. Here, we present the crystal structures of PfHT1, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service