Skip to Content
Merck
All Photos(2)

Key Documents

34102

Supelco

Dicamba methyl ester

PESTANAL®, analytical standard

Synonym(s):

Methyl 3,6-dichloro-2-methoxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8Cl2O3
CAS Number:
Molecular Weight:
235.06
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)c1c(Cl)ccc(Cl)c1OC

InChI

1S/C9H8Cl2O3/c1-13-8-6(11)4-3-5(10)7(8)9(12)14-2/h3-4H,1-2H3

InChI key

AWSBKDYHGOOSML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dicamba methyl ester is a chlorinated acidic herbicide, which exists as a persistent contaminant in the environment.

Application

Dicamba methyl ester may be used as an analytical reference standard for the estimation of the analyte in aqueous samples using chemical derivatization combined with solid-phase extraction, followed by gas chromatography (SPE-GC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of chlorinated phenoxy acid and ester herbicides in soil and water by liquid chromatography particle beam mass spectrometry and ultraviolet absorption spectrophotometry.
Kim SI, et al.
Analytical Chemistry, 63(8), 819-823 (1991)
Establishing the feasibility of coupled solid-phase extraction--solid-phase derivatization for acidic herbicides.
Yu ZL and Wells MJM
Journal of Chromatography A, 1143(1-2), 16-25 (2007)
Anabel Fernández-Iglesias et al.
Journal of cellular and molecular medicine, 23(2), 877-886 (2018-11-13)
Liver cells isolated from pre-clinical models are essential tools for studying liver (patho)physiology, and also for screening new therapeutic options. We aimed at developing a new antibody-free isolation method able to obtain the four main hepatic cell types (hepatocytes, liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service