Skip to Content
Merck
All Photos(1)

Key Documents

30310

Sigma-Aldrich

Cytidine sulfate salt

puriss., crystallized, ≥99.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O5 · 1.5H2SO4
CAS Number:
Molecular Weight:
390.33
Beilstein:
3825931
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

puriss.

Assay

≥99.0% (T)

quality

crystallized

mp

~230 °C (dec.)

SMILES string

OS(O)(=O)=O.NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O.NC3=NC(=O)N(C=C3)[C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service