25264
4-(Chloromethyl)pyridine hydrochloride
purum, ≥97.0% (AT)
Synonym(s):
4-Picolyl chloride hydrochloride
About This Item
grade
purum
Assay
≥97.0% (AT)
mp
166-173 °C (lit.)
storage temp.
2-8°C
SMILES string
Cl[H].ClCc1ccncc1
InChI
1S/C6H6ClN.ClH/c7-5-6-1-3-8-4-2-6;/h1-4H,5H2;1H
InChI key
ZDHKVKPZQKYREU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service