Skip to Content
Merck
All Photos(1)

Key Documents

24981

Sigma-Aldrich

Dimethyl chloromalonate

technical, ~95% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH(COOCH3)2
CAS Number:
Molecular Weight:
166.56
Beilstein:
1768415
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

technical

Assay

~95% (GC)

impurities

~5% dimethyl dichloromalonate

refractive index

n20/D 1.437 (lit.)
n20/D 1.437

bp

105-106 °C/19 mmHg (lit.)

density

1.305 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(Cl)C(=O)OC

InChI

1S/C5H7ClO4/c1-9-4(7)3(6)5(8)10-2/h3H,1-2H3

InChI key

LNBQBURECUEBKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service