Skip to Content
Merck
All Photos(1)

Key Documents

24475

Sigma-Aldrich

2-Chloro-3,5-dinitropyridine

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2ClN3O4
CAS Number:
Molecular Weight:
203.54
Beilstein:
196026
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

≥98.0% (HPLC)

mp

63-65 °C (lit.)
63-65 °C

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cnc(Cl)c(c1)[N+]([O-])=O

InChI

1S/C5H2ClN3O4/c6-5-4(9(12)13)1-3(2-7-5)8(10)11/h1-2H

InChI key

QLHVJBXAQWPEDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Terminal N-blocking reagent for proteins; Condensing agent for esterifying hindered esters

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Takimoto et al.
Bulletin of the Chemical Society of Japan, 56, 639-639 (1983)
A. Signor et al.
Gazzetta Chimica Italiana, 93, 65-65 (1963)
Gazzetta Chimica Italiana, 94, 619-619 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service