Skip to Content
Merck
All Photos(1)

Key Documents

22709

Sigma-Aldrich

Quinuclidine

purum, ≥97.0% (NT)

Synonym(s):

1-Azabicyclo[2.2.2]octane, ABCO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13N
CAS Number:
Molecular Weight:
111.18
Beilstein:
103111
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

1.5 mmHg ( 20 °C)

grade

purum

Assay

≥97.0% (NT)

mp

155-160 °C
157-160 °C (lit.)

SMILES string

C1CN2CCC1CC2

InChI

1S/C7H13N/c1-4-8-5-2-7(1)3-6-8/h7H,1-6H2

InChI key

SBYHFKPVCBCYGV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Werner J Geldenhuys et al.
Bioorganic & medicinal chemistry letters, 20(16), 4870-4877 (2010-07-20)
The high affinity neuronal choline transporter (CHT1) is responsible for the uptake of choline into the pre-synaptic terminal of cholinergic neurons. Considering our past experience with modeling the blood-brain barrier choline transporter (BBBCHT) as drug delivery vector to the CNS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service