Skip to Content
Merck
All Photos(1)

Key Documents

17036

Millipore

N′-Benzoyl-N,N-diisobutylthiourea

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2OS
CAS Number:
Molecular Weight:
292.44
Beilstein:
3336497
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99.0% (HPLC)
≥99.0%

mp

134 °C

SMILES string

CC(C)CN(CC(C)C)C(=S)NC(=O)c1ccccc1

Other Notes

Modifier for preconcentration of silver ions in voltammetric determination[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Guttmann et al.
Analytical and bioanalytical chemistry, 356(3-4), 263-266 (1996-09-01)
The preconcentration of silver at carbon paste electrodes (CPE) modified with N-benzoyl-N',N'-di-i-butyl-thiourea and its subsequent voltammetric determination is studied with test solutions. The preconcentration of silver at the surface of the modified CPE succeeds as well with open circuits as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service