Skip to Content
Merck
All Photos(1)

Key Documents

16718

Supelco

N-Phenyl-α-(4-nitrophenyl)nitrone

Selectophore, ≥97.0%

Synonym(s):

α-(4-Nitrophenyl)-N-phenylnitrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2O3
CAS Number:
Molecular Weight:
242.23
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Assay

≥97.0% (HPLC)
≥97.0%

technique(s)

titration: suitable

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(cc1)\C=[N+](/[O-])c2ccccc2

InChI

1S/C13H10N2O3/c16-14(12-4-2-1-3-5-12)10-11-6-8-13(9-7-11)15(17)18/h1-10H/b14-10-

InChI key

UVECRWKWUJOHSR-UVTDQMKNSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Indicator electrode for the potentiometric titration of Cu2+ with EDTA

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.M. Ardakani et al.
New. J. Chem., 27, 1140 -1140 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service