15575
9-Borabicyclo[3.3.1]nonyl trifluoromethanesulfonate solution
purum, ~0.5 M in hexane
Synonym(s):
9-BBN triflate solution, 9-BBN·CF3SO3, 9-[[(Trifluoromethyl)sulfonyl]oxy]-9-borabicyclo[3.3.1]nonane
About This Item
Recommended Products
grade
purum
reaction suitability
reagent type: reductant
concentration
~0.5 M in hexane
SMILES string
FC(F)(F)S(=O)(=O)OB1C2CCCC1CCC2
InChI
1S/C9H14BF3O3S/c11-9(12,13)17(14,15)16-10-7-3-1-4-8(10)6-2-5-7/h7-8H,1-6H2/t7-,8+
InChI key
NXHUKDCGEGEZCX-OCAPTIKFSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Preparation of a dialkylborenium ion stabilized by an N-heterocyclic carbene
- Mukaiyama aldol addition
- Preparation of porphyrin dimethyldioxanyl tolyl derivative complex
- Preparation of palladium porphyrin complexes
- Boron-mediated β-alkoxy Me ketone aldol addition reactions
- Enolization agent
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service