Skip to Content
Merck
All Photos(1)

Key Documents

12766

Sigma-Aldrich

Benzopurpurine 4B

indicator (pH 2.3-4.4), for microscopy (Hist., Vit.)

Synonym(s):

Direct red 2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H26N6Na2O6S2
CAS Number:
Molecular Weight:
724.72
Colour Index Number:
23500
EC Number:
MDL number:
UNSPSC Code:
12171500
Pricing and availability is not currently available.

grade

for microscopy (Hist., Vit.)
indicator (pH 2.3-4.4)

SMILES string

[Na+].[Na+].Cc1cc(ccc1\N=N\c2cc(c3ccccc3c2N)S([O-])(=O)=O)-c4ccc(\N=N\c5cc(c6ccccc6c5N)S([O-])(=O)=O)c(C)c4


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service