Skip to Content
Merck
All Photos(1)

Key Documents

12278

Sigma-Aldrich

Chromium(III) nitrate solution

64-66% Cr(NO3)3*9H2O basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cr(NO3)3
CAS Number:
Molecular Weight:
238.01
MDL number:
UNSPSC Code:
12352103

Assay

64-66% Cr(NO3)3*9H2O basis

pH

1-3 (20 °C, 5%)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤300 mg/kg

cation traces

Ca: ≤1000 mg/kg
Fe: ≤1000 mg/kg
Na: ≤1000 mg/kg

SMILES string

[Cr+3]

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Ox. Liq. 3 - Skin Corr. 1 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service