Skip to Content
Merck
All Photos(1)

Key Documents

03739

Sigma-Aldrich

2,2′-(Ethylenedioxy)bis(ethylamine)

purum, ≥96.0% (GC)

Synonym(s):

1,2-Bis(2-aminoethoxy)ethane, 1,8-Diamino-3,6-dioxaoctane, 2,2′-(Ethylenedioxy)diethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OCH2CH2OCH2CH2NH2
CAS Number:
Molecular Weight:
148.20
Beilstein:
1739187
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

purum

Assay

≥96.0% (GC)

impurities

≤1% water

refractive index

n20/D 1.461 (lit.)
n20/D 1.461

bp

105-109 °C/6 mmHg (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

NCCOCCOCCN

InChI

1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2

InChI key

IWBOPFCKHIJFMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Li et al.
Nanomedicine : nanotechnology, biology, and medicine, 10(7), 1465-1475 (2014-02-04)
Carbon nanotubes (CNTs) are promising drug delivery systems due to their external functionalizable surface and their hollowed cavity that can encapsulate several bioactive molecules. In this study, the chemotherapeutic drug cisplatin or an inert platinum(IV) complex were entrapped inside functionalized-multi-walled-CNTs
Vinitha Ganesan et al.
Biochimica et biophysica acta, 1854(6), 592-600 (2014-12-04)
Immunoprecipitation (IP) is a widely used technique for identifying the binding partners of the target proteins of specific antibodies. Putative binding targets and their partners are usually in much lower amounts than the antibodies used to capture these target proteins.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service