Skip to Content
Merck
All Photos(1)

Documents

01433

Millipore

Biochemical Esculin ID Membrane

suitable for microbiology, light brown colored, sterile, diameter 70 mm

Synonym(s):

Esculin Test ID Membrane, Indicator membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

agriculture
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Streptococcus spp.

General description

Biochemical Esculin ID Membranes are substrate-carrying membranes used for rapid identification and differentiation of Group D Streptococci through characteristic color reactions. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The group D Streptococci are differentiated from non-group D Streptococci by the detection of esculin hydrolysis. Group D Streptococci hydrolyze the esculin to esculetin and dextrose. Esculetin reacts with an iron salt such as ferric citrate to form a blackish-brown colored complex. The reaction runs on a membrane that gets into contact with colony material.

Application

Biochemical Esculin ID Membrane is recommended to detect Group D Streptococci from pharmaceutical preparations, raw materials, cosmetic samples, etc. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Escherichia coli (25922) - ColorlessEnterococcus faecalis (29212) - BlackEnterococcus faecium (27273) - BlackStaphylococcus aureus (25923) - Colorless

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service