15-2120
L-Isoleucine
SAJ special grade, ≥99.0%
Synonym(s):
(2S,3S)-2-Amino-3-methylpentanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C2H5CH(CH3)CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721792
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
grade
SAJ special grade
Assay
≥99.0%
form
solid
availability
available only in Japan
mp
288 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
CC[C@H](C)[C@H](N)C(O)=O
InChI
1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1
InChI key
AGPKZVBTJJNPAG-WHFBIAKZSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Urho M Kujala et al.
Circulation, 127(3), 340-348 (2012-12-22)
Long-term physical inactivity seems to cause many health problems. We studied whether persistent physical activity compared with inactivity has a global effect on serum metabolome toward reduced cardiometabolic disease risk. Sixteen same-sex twin pairs (mean age, 60 years) were selected
Anna L Vestergaard et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(14), E1334-E1343 (2014-04-08)
P4-ATPases (flippases) translocate specific phospholipids such as phosphatidylserine from the exoplasmic leaflet of the cell membrane to the cytosolic leaflet, upholding an essential membrane asymmetry. The mechanism of flipping this giant substrate has remained an enigma. We have investigated the
Eva Blomstrand et al.
The Journal of nutrition, 136(1 Suppl), 269S-273S (2005-12-21)
BCAAs (leucine, isoleucine, and valine), particularly leucine, have anabolic effects on protein metabolism by increasing the rate of protein synthesis and decreasing the rate of protein degradation in resting human muscle. Also, during recovery from endurance exercise, BCAAs were found
L Edvinsson
Blood vessels, 28(1-3), 35-45 (1991-01-01)
The cerebral circulation is supplied with two vasodilator systems: the parasympathetic system storing vasoactive intestinal peptide, peptide histidine isoleucine, acetylcholine and in a subpopulation of nerves neuropeptide Y, and the sensory system, mainly originating in the trigeminal ganglion, storing substance
Toshihiro Sato et al.
The Journal of biological chemistry, 278(6), 4314-4321 (2002-11-22)
Previously, we determined the crystal structures of the dimeric ligand binding region of the metabotropic glutamate receptor subtype 1. Each protomer binds l-glutamate within the crevice between the LB1 and LB2 domains. We proposed that the two different conformations of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service