Skip to Content
Merck
All Photos(1)

Documents

01-4870

Sigma-Aldrich

Ammonium oxalate monohydrate

JIS special grade, ≥99.5%

Synonym(s):

Oxalic acid diammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2C2O4 · H2O
CAS Number:
Molecular Weight:
142.11
Beilstein:
3759830
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.5%

form

crystalline

availability

available only in Japan

pH

6.0-7.0 (25 °C, 14.2 g/L)

mp

133 °C (dec.) (lit.)

SMILES string

OC(C(O)=O)=O.N.O.N

InChI

1S/C2H2O4.2H3N.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);2*1H3;1H2

InChI key

MSMNVXKYCPHLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Sato
Phytochemistry, 19, 1939-1939 (1980)
Yuancun Qiao et al.
The journal of physical chemistry letters, 6(14), 2755-2760 (2015-08-13)
Negative linear compressibility (NLC) is a relatively uncommon phenomenon and rarely studied in organic systems. Here we provide the direct evidence of the persistent NLC in organic mineral ammonium oxalate monohydrate under high pressure using synchrotron X-ray powder diffraction, Raman
Electrochemiluminescence as a tool for microscopy at the nanoscale.
Lee A, et al.
Proceedings of SPIE, 13, 13-20 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service