Skip to Content
Merck
All Photos(1)

Documents

01-4550

Sigma-Aldrich

Ammonium chloride

JIS special grade, ≥99.0%

Synonym(s):

Salmiac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Beilstein:
4371014
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

vapor density

1.9 (vs air)

vapor pressure

1 mmHg ( 160.4 °C)

Assay

≥99.0%

form

crystalline powder

availability

available only in Japan

pH

4.5-5.5 (20 °C, 50 g/L)

mp

340 °C (subl.) (lit.)

SMILES string

N.Cl

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander J F Egan et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(22), 8197-8202 (2014-05-14)
Bacteria surround their cytoplasmic membrane with an essential, stress-bearing peptidoglycan (PG) layer. Growing and dividing cells expand their PG layer by using membrane-anchored PG synthases, which are guided by dynamic cytoskeletal elements. In Escherichia coli, growth of the mainly single-layered
Allison L Boyd et al.
The Journal of experimental medicine, 211(10), 1925-1935 (2014-09-03)
Allogeneic hematopoietic stem cell (HSC) transplantation (HSCT) is currently the leading strategy to manage acute myeloid leukemia (AML). However, treatment-related morbidity limits the patient generalizability of HSCT use, and the survival of leukemic stem cells (LSCs) within protective areas of
Christine Burkard et al.
PloS one, 9(7), e101762-e101762 (2014-07-16)
Studies of viral entry into host cells often rely on the detection of post-entry parameters, such as viral replication or the expression of a reporter gene, rather than on measuring entry per se. The lack of assays to easily detect
Cyrill A Rentsch et al.
European urology, 66(4), 677-688 (2014-03-29)
Whether the commonly used bacillus Calmette-Guérin (BCG) strains Connaught and Tice confer different treatment responses in non-muscle-invasive bladder cancer (NMIBC) is unknown. To compare clinical efficacy, immunogenicity, and genetics of BCG Connaught and Tice. A prospective randomized single-institution trial with
Sewa Rijal et al.
Blood, 125(18), 2815-2824 (2015-03-05)
Phosphoinositide signaling regulates diverse cellular functions. Phosphoinositide-3 kinase (PI3K) generates PtdIns(3,4,5)P3 and PtdIns(3,4)P2, leading to the activation of proliferative and anti-apoptotic signaling pathways. Termination of phosphoinositide signaling requires hydrolysis of inositol ring phosphate groups through the actions of PtdIns(3,4,5)P3 3-phosphatase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service