Skip to Content
Merck
All Photos(1)

Documents

SQPAKBI0M1

Milli-Q

SQPAK Bio Final Filter

For pyrogen, nuclease, protease and bacteria-free ultrapure water. Use with ultrapure dispensing module of Milli-Q® SQ 2Series systems.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190

Quality Level

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® SQ 2Series

General description

To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
The SQPAK Bio Final filter ensures high-quality ultrapure (Type 1) water at the point of dispense.
The purification technology includes an ultrafiltration filter that removes bacteria, particulates and macromolecules (such as pyrogens, nucleases and proteases) from ultrapure water.

For use with the ultrapure dispensing module of Milli-Q® SQ 2Series configurations, such as Milli-Q® SQ 200/200P and Milli-Q® SQ 240C/240CV systems.

Application

The SQPAK Bio Final filter is useful for lab applications involving cell and molecular biology.

Features and Benefits

  • Easy replacement by simply twisting
  • Replacement is recommended after 3 months

Other Notes

Directions For Use:
  • Organism Retention: Bacteria and pyrogens
  • Mode of Action: Ultrafiltration
  • Application: Cell culture media, molecular biology
  • Intended Use: Bioburden reduction
  • Instructions for Use: This item provides water filtered through ultrafiltration hollow fibers.
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SQPAK is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.

Related Content

Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service