SCC443
12Z Human Endometrial Epithelial Cell Line
Synonym(s):
endometrial cell line, endometriosis cell line, 12Z
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Recommended Products
biological source
human
Quality Level
packaging
vial of ≥1X10⁶ cells/vial vial
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
cell culture | mammalian: suitable
shipped in
liquid nitrogen
storage temp.
−196°C
General description
Endometriosis is a gynecologic condition that is characterized by the presence of endometrial tissue outside of the uterus. This condition affects approximately 6-10% of all women and can lead to significant pelvic pain and infertility. The limited availability of in vitro models hinders cellular, molecular, and pathophysiologic studies of endometriosis which translates to poor understanding of the disease and limited available treatments.
Several endometriotic cell lines were developed by researchers from Goethe University Frankfurt in Germany. One of these is the 12Z cell line which was derived from the endometrial peritoneum of a 37 year-old female undergoing laparoscopy. The 12Z cell line was immortalized using SV40 plasmid transfection. This cell line expresses both pan-cytokeratin and N-Cadherin.
One of the frequently-studied means for the initiation of endometriosis is retrograde menstruation that permits the deposition of endometrial tissues outside of the uterus. During this process, the invasive capabiity of cells is believed to be essential.
The 12Z immortalized human endometrial epithelial cell line is an established model for endometriosis as it is derived from endometriotic tissue that is positive for N-Cadherin+, a marker for invasiveness. This makes the 12Z iine a suitable model for the study of disease progression or therapeutic development. The 12Z cell line is frequently used in 2D or 3D invasion assays, as well as in biochemical assays to study the pathophysiology of endometriosis.
Source
The 12Z cell line was derived from endometrial peritoneum biopsy of a 37 year-old female. This cell line was immortalized via transfection of a plasmid containing SV40T.
References
1. Nat. Rev. Endocrinol 2019, 15(11): 666-682.
2. Lancet 2004, 364 (9447): 1789-1799
3. Am. J. Pathol 2001, 159(5): 1839-1852
4. Reproductive BioMedicine Online 2022, 44(6): 976-990. DOI: https://doi.org/10.1016/j.rbmo.2022.02.008
5. Sci Rep 2021, 11(1): 4115
Several endometriotic cell lines were developed by researchers from Goethe University Frankfurt in Germany. One of these is the 12Z cell line which was derived from the endometrial peritoneum of a 37 year-old female undergoing laparoscopy. The 12Z cell line was immortalized using SV40 plasmid transfection. This cell line expresses both pan-cytokeratin and N-Cadherin.
One of the frequently-studied means for the initiation of endometriosis is retrograde menstruation that permits the deposition of endometrial tissues outside of the uterus. During this process, the invasive capabiity of cells is believed to be essential.
The 12Z immortalized human endometrial epithelial cell line is an established model for endometriosis as it is derived from endometriotic tissue that is positive for N-Cadherin+, a marker for invasiveness. This makes the 12Z iine a suitable model for the study of disease progression or therapeutic development. The 12Z cell line is frequently used in 2D or 3D invasion assays, as well as in biochemical assays to study the pathophysiology of endometriosis.
Source
The 12Z cell line was derived from endometrial peritoneum biopsy of a 37 year-old female. This cell line was immortalized via transfection of a plasmid containing SV40T.
References
1. Nat. Rev. Endocrinol 2019, 15(11): 666-682.
2. Lancet 2004, 364 (9447): 1789-1799
3. Am. J. Pathol 2001, 159(5): 1839-1852
4. Reproductive BioMedicine Online 2022, 44(6): 976-990. DOI: https://doi.org/10.1016/j.rbmo.2022.02.008
5. Sci Rep 2021, 11(1): 4115
Application
Each vial contains >=1X106 viable cells.
Cells are tested negative for infectious diseases by a Human Essential CLEAR Panel by Charles River Animal Diagnostic Services.
Cells are verified to be of human origin and negative for interspecies contamination from rat, mouse, Chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
Cells are negative for mycoplasma contamination.
Cells are tested negative for infectious diseases by a Human Essential CLEAR Panel by Charles River Animal Diagnostic Services.
Cells are verified to be of human origin and negative for interspecies contamination from rat, mouse, Chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
Cells are negative for mycoplasma contamination.
Features and Benefits
The N-cadherin-positive 12Z human endometrial cell line is an established model for tissue invasion in endometriosis, and is used to study disease progression, pathogenesis, and therapeutic intervention.
Storage and Stability
Store cells in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and function.
Other Notes
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unlessotherwise stated in our catalog or other company documentation accompanying theproduct(s), our products are intended for research use only and are not to beused for any other purpose, which includes but is not limited to, unauthorizedcommercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic usesor any type of consumption or application to humans oranimals.;This product contains genetically modified organisms(GMO). Within the EU GMOs are regulated by Directives 2001/18/ECand 2009/41/EC of the European Parliament and of the Council and their nationalimplementation in the member States respectively. This legislation obliges<{HCompany}/> to request certain information about you and theestablishment where the GMOs are being handled. Click here for EnduserDeclaration (EUD) Form.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service