Progard® TS2 Pretreatment Pack is not classified in Europe as a hazardous substance in regards to the substance definition in Article 3 of the Regulation (EC) 1907/2006 of 18 December 2006 concerning the Registration, Evaluation, Authorization and Restriction of Chemical (REACH) and in regards to the hazardous substance and mixtures classification criteria from the Annex I of the Regulation (EC) 1272/2008 of 16 December 2008 on classification, labeling and packaging of substance and mixtures.
Therefore, this Purification Cartridge do not require in Europe any Safety Data Sheet (SDS) to be in accordance with the Regulation (EC) 1907/2006 and the Regulation (EC) 1272/2008.
Progard® TS2 Pretreatment Pack do not require a Safety Data Sheet (SDS) under the Occupational Health and Safety Administration Standard entitled “Hazard Communication” 29 CFR 1910.1200 for the United States of America.
Progard® TS2 Pretreatment Pack do not meet the criteria of a W.H.M.I.S. classification of a controlled product. As a result, a W.H.M.I.S. Material Safety Data Sheet is not required in Canada for this product.
Disposable of wastes should be in accordance with applicable regional, national, and local laws and regulations.
PR0G0T0S2
Progard® TS2 Pretreatment Pack
For the optimal protection of Reverse Osmosis (RO) membranes
Select a Size
Select a Size
About This Item
Recommended Products
material
EPDM rubber
cartridge (activated carbon)
cartridge
polypropylene
Quality Level
feature
Optimal protection of Reverse Osmosis (RO) membrane, optimized system lifetime and performance. Removes of large particles and free chlorine from feed water with a silt density index (SDI) up to 12.
packaging
pkg of 1 unit
H
39.5 cm (15.5 in.)
compatibility
for use with AFS®
for use with Elix® Advantage
for use with Elix® Essential UV
for use with Elix® Essential
for use with Elix® Reference
for use with Milli-Q® Integral
for use with RiOs™ Essential
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Protects RO membrane against mineral scaling, organic fouling and chlorine oxidation.
- Polyphosphates provide protection against scaling.
- Silver-impregnated activated carbon reduces free chlorine and prevents bacteria proliferation.
- Depth-filtration removes particles and colloids.
- e-Sure RFID tag ensures full traceability and provides instant visibility of pack parameters.
- Replace every 6 months for optimal results.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Come smaltire i blocchi già utilizzati?
1 answer-
Helpful?
-
-
What are the storage conditions for this product?
1 answer-
Recommended storage conditions: temperatures from 5 to 30°C; humidity between 10 and 80%. The original sealed bag (if provided) must be kept intact before the first use to protect the purification element from foreign matter.
Helpful?
-
-
How are consumables changed?
1 answer-
To help you to change your system's consumables, several video tutorials are available on our YouTube channel: https://youtube.com/playlist?list=PL2TUriaAaEA_IXDCnPbDk6OmrwptjwOHD. For high-flow systems, please keep in mind that all consumable changes must be approved by a 3-digit code that only the person in charge of the system knows.
Helpful?
-
-
How can I know which consumables can be used with my system?
1 answer-
To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.
Helpful?
-
-
What steps are you taking towards improved sustainability?
1 answer-
Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.
Helpful?
-
-
Can I arrange to have my consumables automatically ordered and delivered once a year?
1 answer-
It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).
Helpful?
-
-
Can consumables be opened and refilled?
1 answer-
Unfortunately, this is not possible because of safety issues and the need to guarantee the quality of consumables and their interior purification components. Our consumables are manufactured in an ISO 9001-certified facility France in agreement with high quality standards.
Helpful?
-
-
What is the consumables delivery time?
1 answer-
Refer to the consumable's Product page for a first estimation of its expected delivery time and contact customer service for more information.
Helpful?
-
-
How frequently do the various consumables need to be changed?
1 answer-
The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.
Helpful?
-
-
Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?
1 answer-
Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service