Skip to Content
Merck
All Photos(1)

Key Documents

MABX8405

Sigma-Aldrich

Anti-SARS-CoV-2 S humanized antibody (2C1)

suitable for ELISA

Synonym(s):

2019-nCoV, SARS-CoV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in human cells

Quality Level

antibody form

culture supernatant

clone

2C1, monoclonal

storage condition

avoid repeated freeze/thaw cycles

technique(s)

ELISA: suitable

shipped in

dry ice

General description

This human recombinant monoclonal Anti-SARS-CoV-2 S humanized antibody (2C1), Cat. No. MABX8405 is tested for use in ELISA, for the detection of SARS-CoV-2 Spike.

Specificity

SARS CoV2

Application

Recommend MABX8403 for paired antibody immunoassay.

Target description

Coronavirus disease 2019 (COVID-19) is an infectious disease caused by the virus strain severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus closely related to the SARS virus. A SARS-CoV-2 spike protein antibody can be used to support the detection of SARS-CoV-2 (COVID-19, 2019-nCoV).

Physical form

Purified human recombinant monoclonal in buffer containing 20mM Tris and 50mM Glycine without preservatives.

Reconstitution

Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service