MABX51
Anti-Chikungunya (CHIKV-Env 1)
Synonym(s):
Anti-CHIKV Antibody
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
clone
monoclonal
form
solution
species reactivity
all
packaging
pkg of 1 mg (MABX51-1KC)
pkg of 10 mg (MABX51-10KC)
technique(s)
ELISA: suitable
immunodiffusion: suitable
isotype
IgG2a
relevant disease(s)
infectious diseases
shipped in
wet ice
storage temp.
2-8°C
Specificity
Chikungunya Virus Envelope 1 Protein. No reactivity with Dengue and Zika Viruses
Immunogen
Recombinant Chikungunya Virus Envelope Protein
Application
This mouse monoclonal antibody, Cat. No. MABX51, is tested for use in Lateral Flow Assay and ELISA, for the detection of Recombinant Chikungunya Virus Envelope 1 Protein.
Target description
Chikungunya virus is an arthropod-borne virus (arbovirus) spread by the bite of infected Aedes mosquitoes. It is a member of the Alphavirus genus of RNA viruses. Symptoms of fever and joint pain usually appear within a week of infection. Muscle pain, headache, fatigue and rash may also occur.
Linkage
Recommended Matched Pair MABX51 with MABX49.
Physical form
Purified mouse monoclonal in buffer containing 0.015 M Potassium Phosphate, pH 7.2 with 0.85% Sodium Chloride and 0.05% Sodium Azide
Reconstitution
Please refer to Certificate of Analysis for concentration.
Storage and Stability
Store at 2-8 °C. For long-term storage, freeze at -20 °C
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service