Skip to Content
Merck
All Photos(2)

Documents

MABT356

Sigma-Aldrich

Anti-Integrin alpha6 Antibody, clone 6H7.1

clone 6H7.1, from mouse

Synonym(s):

Integrin alpha-6, CD49 antigen-like family member F, VLA-6, CD49f

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6H7.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA6(3655)

General description

Integrin alpha-6 (UniProt P23229; also known as CD49 antigen-like family member F, CD49f, Integrin alpha6B, VLA-6) is encoded by the ITGA6 (also known as ITGA6B) gene (Gene ID 3655) in human. The biological functions attributed to cell-extracellular matrix (ECM) interactions are mediated primarily by integrin family transmembrane receptors. A total of 18 alpha and 8 beta integrins have been identified in human. Each alpha integrin pairs up with one beta integrin non-covalently to form a functional heterodimeric dimer. At least 24 distinct integrin heterodimers are known with varying ligand specificities. For example, the alpha6-beta1 integrin is reported to mediate the spreading and laminin-binding of Schwann cells, while the alpha6-beta4 integrin is shown to play a role in the suppression of anoikis in human intestinal epithelial cells (IECs).

Signal (1-23), Integrin alpha-6 (24-1130). Processed into Integrin alpha-6 heavy chain (24 – 938), Integrin alpha-6 light chain (942-1130), Processed integrin alpha-6 (Alpha6p; 636 – 1130)

Specificity

Expected to react with Integrin alpha-6 spliced isoforms 2 (Alpha-6X1A), 4 (Alpha-6X2A ), 6 (Alpha-6X1X2A), 7, and 9, as well as their respective proforms, cleaved light chains, and processed integrin alpha-6 forms (Alpha6p). Epitope is not present in isoform 1 (Alpha-6X1X2B), 3 (Alpha-6X1B), or 5 (Alpha-6X2B).

Immunogen

Epitope: Cytoplasmic region of isoforms 2/4/6/7/9
KLH-conjugated linear peptide corresponding to the cytoplasmic region of human Integrin alpha6 isoforms 2/4/6/7/9.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Integrin alpha6 in human skin tissue.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Integrin alpha6 Antibody, clone 6H7.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of Integrin alpha6.

Quality

Evaluated by Western Blotting in SW480 human colon cancer cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Integrin alpha6 in 10 µg of SW480 human colon cancer cell lysate.

Target description

~140 kDa observed. Due to glycosylation, the target band(s) appears larger than the calculated 106.6 to 124.0 kDa based on the proform sequences of isoforms 2/4/6/7/9. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carina Magdaleno et al.
Journal of cellular biochemistry, 122(5), 524-537 (2021-01-14)
Fibronectin (FN) is an extracellular matrix (ECM) glycoprotein that self-assembles into FN fibrils, forming a FN matrix contributing to the stiffness of the ECM. Stromal FN stiffness in cancer has been shown to impact epithelial functions such as migration, cancer
Seo-Hyun Choi et al.
Molecular cancer research : MCR, 20(8), 1305-1319 (2022-04-09)
KRAS mutation in colorectal cancer is associated with aggressive tumor behavior through increased invasiveness and higher rates of lung metastases, but the biological mechanisms behind these features are not fully understood. In this study, we show that KRAS-mutant colorectal cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service