Skip to Content
Merck
All Photos(2)

Documents

MABS1822

Sigma-Aldrich

Anti-SHMT Antibody, clone 4D2.1

clone 4D2.1, from mouse

Synonym(s):

Serine hydroxymethyltransferase mitochondrial, EC: 2.1.2.1, SHMT, Glycine hydroxymethyltransferase, Serine methylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D2.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SHMT2(6472)

General description

Serine hydroxymethyltransferase, mitochondrial (UniProt: P34897; also known as EC: 2.1.2.1, SHMT, Glycine hydroxymethyltransferase, Serine methylase) is encoded by the SHMT2 gene (Gene ID: 6472) in human. SHMT is involved in the de novo mitochondrial thymidylate biosynthesis pathway via its role in glycine and tetrahydrofolate metabolism. Thymidylate biosynthesis is required to prevent uracil accumulation in mtDNA. SHMT catalyzes the reversible conversion of serine and glycine. It uses pyridoxal-5 -phosphate as a cofactor. SHMT is synthesized with a transit peptide (aa 1-29) that is needed for its mitochondrial translocation. In eukaryotes two forms of SHMT have been reported: cytosolic and mitochondrial. In the presence of bound pyridoxal 5′-phosphate it is present in a homotetrameric form and in the absence of pyridoxal 5′-phosphate it exists as a homodimer. Pyridoxal 5′-phosphate binding mediates an important conformation change, which is essential for its tetramerization. Three isoforms of SHMT have been reported that are produced by alternative splicing.

Specificity

Clone 4D2.1 detects human mitochondrial Serine hydroxymethyltransferase. It targets an epitope within 107 amino acids from the C-terminal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 107 amino acids from the C-terminal half of human mitochondrial Serine hydroxymethyltransferase.

Application

Anti-SHMT, clone 4D2.1, Cat. No. MABS1822, is a mouse monoclonal antibody that detects mitochondrial Serine hydroxymethyltransferase and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50-250 dilution of this antibody from a representative lot detected SHMT in human colon, human liver, and human pancreas tissue sections.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SHMT in Jurkat cell lysate.
Research Category
Signaling

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SHMT in Raji cell lysate.

Target description

~56 kDa observed; 55.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service