MABN85
Anti-CORO1A Antibody, clone 2E11.2
clone 2E11.2, from mouse
Synonym(s):
Coronin-1A, Coronin-like protein A, Clipin-A, Coronin-like protein p57, Tryptophan aspartate-containing coat protein, TACO
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2E11.2, monoclonal
species reactivity
mouse, human
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Coro1A(12721)
General description
Immunogen
Application
Neuroscience
Developmental Signaling
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected CORO1A in human cerebral cortex issue.
Quality
Western Blotting Analysis: 2.0 µg/mL of this antibody detected CORO1A in 10 µg of Raji cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service