Skip to Content
Merck
All Photos(3)

Key Documents

MABF249

Sigma-Aldrich

Anti-Tapasin Antibody, clone 7F6

clone 7F6, from rat

Synonym(s):

Tapasin, TPN, TSPN, NGS-17, TAP-associated protein, TAP-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7F6, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TAPBP(6892)

Related Categories

General description

Tapasin (TPN or TPSN) is also known as NGS-17, TAP-associated protein and TAP-binding protein. Tapasin plays a role in MHC class I peptide loading and assembly, and is involved in associating MHC class I with transporter associated with antigen processing (TAP). Tapasin is found in neutrophils, predominantly within fully differentiated cells.

Specificity

This antibody recognizes the Lumenal domain of Tapasin.

Immunogen

Linear peptide corresponding to human Tapasin.

Application

Detect Tapasin using this rat monoclonal antibody, Anti-Tapasin Antibody, clone 7F6 validated for use in western blotting & Flow Cytometry.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Tapasin in 10 µg of A431, HEK293, HepG2, HUVEC, Jurkat, and PC12 cell lysates.

Flow Cytometry Analysis: 4 µg from a representative lot detected Tapasin in 1X10E6 PBMC cells.

Western Blotting Analysis: A representative lot from an independent laboratory detected Tapasin in BNLF2aHA transfected MJS cell lysate (Hislop, A. D., et al. (2007). J Exp Med. 204(8):1863-1873.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Tapasin in HeLa cell lysate (Hulpke, S., et al. (2012). Cell Mol Life Sci. 69(19):3317-3327.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Tapasin in BNLF2aHA4M transfected MJS cell lines (Horst, D., et al. (2009). J Immunol. 182(4):2313-2314.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Tapasin in WB (Koch, J., et al. (2006). FEBS Lett. 580(17):4091-4096.).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Tapasin in 10 µg of HeLa cell lysate.

Target description

~48 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Blees et al.
Scientific reports, 5, 17341-17341 (2015-11-28)
Salt bridges in lipid bilayers play a decisive role in the dynamic assembly and downstream signaling of the natural killer and T-cell receptors. Here, we describe the identification of an inter-subunit salt bridge in the membrane within yet another key

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service