Skip to Content
Merck
All Photos(1)

Key Documents

MABF1411

Sigma-Aldrich

Anti-MHC class II (I-A/I-E), PE, clone M5/114 .15.2 Antibody

clone M5/114.15.2, from rat, PE

Synonym(s):

H-2 class II histocompatibility antigen gamma chain, Ia antigen-associated invariant chain (Ii), MHC class II-associated invariant chain, CD74, MHC class II (I-A/I-E)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₹23,080.00

₹23,080.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
₹23,080.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

₹23,080.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

M5/114.15.2, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Cd74(16149)

General description

The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages, and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also be referred to as M5/114 in the literature.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Anti-MHC class II (I-A/I-E), PE, clone M5/114 .15.2 Antibody is an antibody against MHC class II for use in Flow Cytometry.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.06 μg from a representative lot detected MHC class II (I-A/I-E) in one million C57Bl/6 splenocytes.

Physical form

Purified Rat monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.
Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service