Skip to Content
Merck
All Photos(1)

Key Documents

MABE989

Sigma-Aldrich

Anti-Homeodomain-only Protein Antibody, clone 15C1.1

clone 15C1.1, from mouse

Synonym(s):

Lung cancer-associated Y protein, Homeodomain-only Protein, Not expressed in choriocarcinoma protein 1, Odd homeobox protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
clone:
15C1.1, monoclonal
application:
WB
species reactivity:
mouse
technique(s):
western blot: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15C1.1, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HOPX(84525)

General description

Homeodomain-only protein (UniProt Q9BPY8; also known as Lung cancer-associated Y protein, Not expressed in choriocarcinoma clone 1, Not expressed in choriocarcinoma protein 1, Odd homeobox 1 protein, Odd homeobox protein 1) is encoded by the HOPX (also known as CAMEO, HOD, HOP, LAGY, NECC1, OB1, SMAP31, TOTO) gene (Gene ID 84525) in human. Human and mouse HOPX proteins share 92% identity, they contain homeobox-like domain but lack sequence motif required for DNA binding. HOPX is highly expressed in the developing heart, where its expression is dependent on the cardiac-restricted homeobox protein, Nkx2.5. HOPX physically interacts with serum responsive factor (SRF) and inhibits SRF-dependent transcription by inhibiting SRF binding to DNA and by recruiting histone deacetylase (HDAC) activity. HOPX is reported to play a critical role in the differentiation of various cells such as myoblasts, trophoblasts, keratinocytes, T cells, and lung alveolar cells. HOPX was initially reported as a differentiation marker and is expressed in various normal tissues, and HOPX expression is markedly suppressed in a subset of cancers.

Specificity

Expected to react with spliced isoforms 1, 3, and 4. Reactivity toward isoform 2 has not been determined.

Immunogen

GST-tagged recombinant protein corresponding to human Homeodomain-only Protein.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-Homeodomain-only Protein Antibody, clone 15C1.1 is validated for use in Western Blotting for the detection of Homeodomain-only Protein.

Quality

Evaluated by Western Blotting in mouse lung tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Homeodomain-only Protein in 10 µg of mouse lung tissue lysate.

Target description

~10 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service