Skip to Content
Merck
All Photos(1)

Key Documents

MAB011-A/13

Sigma-Aldrich

Anti-Apolipoprotein A-I Antibody, clone A/13

clone A/13, Chemicon®, from mouse

Synonym(s):

ApoAI

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
₹44,730.00

₹44,730.00


Estimated to ship on08 May 2025Details



Select a Size

Change View
250 μG
₹44,730.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

₹44,730.00


Estimated to ship on08 May 2025Details


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

A/13, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
radioimmunoassay: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APOA1(335)

Specificity

Apolipoprotein A-I. No cross reactivity to Apo A-II or Apo B.

AFFINITY CONSTANT: 8 x 10E9 L/mol

Immunogen

Purified human plasma Apo A-I.

Application

Anti-Apolipoprotein A-I Antibody, clone A/13 detects level of Apolipoprotein A-I & has been published & validated for use in ELISA, RIA.
Competitive RIA or EIA

Optimal working dilutions must be determined by the end user.
Research Category
Metabolism
Research Sub Category
Lipid Metabolism & Weight Regulation

Physical form

Format: Purified
IgG fraction purified from mouse ascites by DEAE column chromatography. In 0.015 M potassium phosphate buffer with 0.15M NaCl, 0.1% sodium azide, pH 7.2.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Do not store below -20°C or in dry ice as precipitation may occur. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service