Skip to Content
Merck
All Photos(1)

Documents

CDRCH12HG

Milli-Q

Reverse Osmosis Cartridge

High Flux RO Cartridge with housing, For use with AFS® systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104214
NACRES:
JA.13

packaging

pkg of 1 unit

impurities

>99% Bacteria Rejection
>99% Organics (MW < 100)
>99% Particulates Rejection
98% Typical Ionic Rejection (Minimum 93%)

compatibility

for use with AFS®

Looking for similar products? Visit Product Comparison Guide

General description

The reverse osmosis (RO) membrane removes 95%-99% of contaminants including ions, particulates, bacteria, and organic molecules larger than 200 Da.

Application

Thanks to its tight pore structure, reverse osmosis removes a large amount of contaminants from water, including ions.

Features and Benefits

High Flux RO Cartridge with housing.

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service