Skip to Content
Merck
All Photos(2)

Key Documents

ABT341

Sigma-Aldrich

Anti-KLHL1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Kelch-like 1 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₹34,180.00

₹34,180.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
₹34,180.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

₹34,180.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KLHL1(57626)

General description

The mammalian Kelch-like 1 (KLHL1) was initially discovered as a homolog to the Drosophila Kelch gene that is highly expressed in several brain tissues. KLHL1 contains one BTB (POZ) domain and six Klech repoeats. KLHL1 may play an important role in organizing the actin cytoskeleton of the brain cells. Mutations associated with KLHL1 are inplicated in spinocerebellar ataxia type 8 (SCA8).

Immunogen

KLH-conjugated linear peptide corresponding to human KLHL1.

Application

Detect Kelch-like 1 using this rabbit polyclonal antibody, Anti-KLHL1 Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: 10 µg/mL from a representative lot detected KLHL1 in rat brain tissue.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected KLHL1 in 15 µg/mL of human brain tissue lysate.

Target description

~100 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service