Skip to Content
Merck
All Photos(2)

Key Documents

ABN212

Sigma-Aldrich

Anti-PRCD Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Progressive rod-cone degeneration protein, PRCD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRCD(768206)

General description

Progressive rod-cone degeneration (PRCD) protein is a single pass membrane protein that is involved in vision. PRCD defects can cause retinitis pigmentosa type 36 (RP36) which typically involves a loss of peripheral and night vision.

Specificity

Other homologies: Canine (92% sequence homology)
This antibody recognizes PRCD near the C-terminus.

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to human PRCD near the C-terminus.

Application

Anti-PRCD Antibody is a highly specific rabbit polyclonal antibody, that targets Progressive rod-cone degeneration protein & has been tested in IHC.
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected PRCD in mouse retina and mouse corneal epithelial cell tissues.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Immunohistochemistry in mouse retina tissue.

Imunnohistochemistry Analysis: A 1:2,000 dilution of this antibody detected PRCD in mouse retina tissue.

Target description

6 kDa calculated

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse retina tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service