ABE1325
Anti-FOXK1 (MNF)
from rabbit, purified by affinity chromatography
Synonym(s):
Forkhead box protein K1, Myocyte nuclear factor, MNF
Select a Size
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... FOXK1(221937)
General description
Specificity
Immunogen
Application
Epigenetics & Nuclear Function
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected FOXK1 (MNF) in human cerebral cortex, human liver, and human testis tissues.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected FOXK1 (MNF) in human cerebral cortex, human liver, and human testis tissues.
Quality
Western Blotting Analysis: 2 µg/mL of this antibody detected FOXK1 (MNF) in HeLa cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service