Skip to Content
Merck
All Photos(1)

Key Documents

AB5701-I

Sigma-Aldrich

Anti-Slit2 Antibody, C-term

from rabbit, purified by affinity chromatography

Synonym(s):

Slit homolog 2 protein, Slit-2, Slit2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₹44,559.99

₹44,559.99


Estimated to ship on25 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
₹44,559.99

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

₹44,559.99


Estimated to ship on25 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SLIT2(9353)

General description

Slit homolog 2 protein (UniProt O94813; also known as Slit-2, Slit2) is encoded by the SLIT2 (also known as SLIL3) gene (Gene ID 9353) in human. Slit proteins are essential for midline guidance in the forebrain where they act as repulsive signal preventing inappropriate midline crossing by axons projecting from the olfactory bulb. Slit-2 acts as molecular guidance cue in cellular migration and its function appears to be mediated by interaction with roundabout homolog receptors. During neural development it is involved in axonal navigation at the ventral midline of the neural tube and projection of axons to different regions. In the developing visual system it appears to function as repellent for retinal ganglion axons by providing a repulsion that directs these axons along their appropriate paths prior to, and after passage through, the optic chiasm. Ref.: Niclou, SP., et al. (2000) J Neurosci. 20; 4962-4974.

Specificity

This polyclonal antibody targets region within the CTCK domain (a.a. 1453-1528) ane will therefore react with uncleaved Slit-2 and cleaved Slit-2 C-product (a.a. 1122-1529), but not the cleaved N-product (a.a. 31-1121).

Immunogen

KLH-conjugated linear peptide corresponding to a sequence within the CTCK domain of human Slit-2.

Application

Anti-Slit2, C-term, Cat. No. AB5701-I, is a highly specific rabbit polyclonal antibody that targets Slit2 and has been tested in Western Blotting.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in U2OS cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Slit-2 in 10 µg of U2OS cell lysate.

Target description

~180 kDa observed. 166.8/166.3/165.8 kDa (isoform 1/2/3 without signal peptide a.a. 1-30) and 44.52 kDa (C-product) calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: AB5701

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service