Skip to Content
Merck
All Photos(1)

Key Documents

902319

Supelco

Wire carrier

for use with wide-necked PE bottles (4 to 10 L)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.51
Pricing and availability is not currently available.

compatibility

for use with wide-necked PE bottles (4 to 10 L)

shipped in

ambient

storage temp.

no temp limit

General description

This wire carrier is specially developed to handle and transport wide-necked PE-bottles of 4l to 10l volume easily and safely.
for wide-necked PE bottles (4 l to 10 l volume)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service