Skip to Content
Merck
All Photos(1)

Documents

901922

Supelco

Reducer (PE)

for S 56x4 to 2" thread (2" coarse to 2" fine thread)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

fitting

for S 56x4 to 2" thread (2" coarse to 2" fine thread)

shipped in

ambient

storage temp.

room temp

Related Categories

General description

Special S40 bottle adapters made of PE provide all possible connection options for safe and reliable daily solvents handling. This adapter is equipped with S56x4 inner thread and G2 / 2" outer thread.
from S 56x4 to 2" thread (2" coarse to 2" fine thread)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service